accessibility menu, dialog, popup

UserName

CAS RN 17292-62-5

NaNHN

CAS RN 17292-62-5

IUPAC Name: sodium cyanoazanide
Synonyms: sodium cyanoazanide
Molecular Weight (g/mol): 64.02
Molecular Formula: CHN2Na
InChi Key: MBEGFNBBAVRKLK-UHFFFAOYSA-N
SMILES: [Na+].[NH-]C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results