missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydrogen Sulfate, TRC
Specifications
Specifications
| Chemical Name or Material | Sodium Hydrogen Sulfate |
| CAS | 7681-38-1 |
| Quantity | 1 g |
| Molecular Formula | NaHSO4 |
| Synonym | Acid Sodium Sulfate, BIF, Bisulfate of Soda, Fanal, Monobasic Sodium Sulfate, Monosodium Hydrogen Sulfate, Monosodium Sulfate, Niter cake, Nitre cake, Poultry Litter Treatment, Printgen N, SBS Pet, Sodium Acid Sulfate, Sodium Bisulfate, Sodium Bisulphate, Sodium Hydrogen Sulfate, Sodium Hydrogen Sulfate (NaHSO4), Sodium Hydrosulfate, WC 00, WC-Klosettreiniger, WC-Perfect, WC-Super, pHase (Sodium Bisulfite) |
| InChI Formula | InChI=1S/Na.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+1;/p-1 |
| SMILES | [Na+].OS(=O)(=O)[O-] |
| IUPAC Name | sodium;hydrogen sulfate |
| Molecular Weight (g/mol) | 120.06 |
| Formula Weight | 119.9493 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?