missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydrogen Selenite, purum p.a, ≥96.0% (RT), Solstice

Product Code. 15606820 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15606820 25 g Poly bottle 25g
15696810 100 g Poly bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15606820 Supplier Honeywell Research Chemicals Supplier No. 7165825G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥96.0% (RT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-82-3
Molecular Formula Na2O3Se
Molecular Weight (g/mol) 172.95
MDL Number MFCD00003525
InChI Key BVTBRVFYZUCAKH-UHFFFAOYSA-L
Synonym sodium hydrogen selenite, sodium hydroselenite, sodium biselenite, selenious acid, monosodium salt, sodium selenite nahseo3, sodium hydrogenselenite, hydrogen sodium selenite, unii-him8pp9m79, sodium hydrogen trioxoselenite, hydrogen sodium selenium oxide
PubChem CID 23669629
IUPAC Name sodium;hydrogen selenite
SMILES [Na+].[Na+].[O-][Se]([O-])=O

Specifications

Quantity 25 g
Linear Formula NaHSeO3
UN Number UN2630
Formula Weight 150.96g/mol
Percent Purity ≥96.0% (RT)
Packaging Poly bottle
Chemical Name or Material Sodium hydrogen selenite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.