missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrogen selenite, 96%

Product Code. 11421900
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11421900 1000 g 1000g
11431900 50 g 50g
11441900 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11421900 Supplier Thermo Scientific Alfa Aesar Supplier No. A12173.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7782-82-3 | Na2O3Se | 172.95 g/mol

Sodium hydrogen selenite is used as a culture media additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hydrogen selenite is used as a culture media additive.

Solubility
Soluble in water.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-82-3
Molecular Formula Na2O3Se
Molecular Weight (g/mol) 172.95
MDL Number MFCD00003525
InChI Key BVTBRVFYZUCAKH-UHFFFAOYSA-L
Synonym sodium hydrogen selenite, sodium hydroselenite, sodium biselenite, selenious acid, monosodium salt, sodium selenite nahseo3, sodium hydrogenselenite, hydrogen sodium selenite, unii-him8pp9m79, sodium hydrogen trioxoselenite, hydrogen sodium selenium oxide
PubChem CID 23669629
SMILES [Na+].[Na+].[O-][Se]([O-])=O

Specifications

Quantity 1000 g
UN Number UN2630
Solubility Information Soluble in water.
IUPAC Name disodium selenite
Formula Weight 150.96
Percent Purity 98%
Sensitivity Hygroscopic
Chemical Name or Material Sodium hydrogen selenite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.