missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrogen selenite, 96%

Product Code. 11431900
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11431900

Brand: Thermo Scientific Alfa Aesar A12173.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7782-82-3 | Na2O3Se | 172.95 g/mol

Sodium hydrogen selenite is used as a culture media additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hydrogen selenite is used as a culture media additive.

Solubility
Soluble in water.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-82-3
Molecular Formula Na2O3Se
Molecular Weight (g/mol) 172.95
MDL Number MFCD00003525
InChI Key BVTBRVFYZUCAKH-UHFFFAOYSA-L
Synonym sodium hydrogen selenite, sodium hydroselenite, sodium biselenite, selenious acid, monosodium salt, sodium selenite nahseo3, sodium hydrogenselenite, hydrogen sodium selenite, unii-him8pp9m79, sodium hydrogen trioxoselenite, hydrogen sodium selenium oxide
PubChem CID 23669629
SMILES [Na+].[Na+].[O-][Se]([O-])=O

Specifications

Quantity 50 g
UN Number UN2630
Solubility Information Soluble in water.
IUPAC Name sodium;hydrogen selenite
Formula Weight 150.96
Percent Purity 98%
Sensitivity Hygroscopic
Chemical Name or Material Sodium hydrogen selenite
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium hydrogen selenite
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H331-Toxic if inhaled.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P314-Get medical advice/attention if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Sodium hydrogen selenite

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.