missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrogen phosphate heptahydrate, ACS, 98.0-102.0%

Product Code. 11386429
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11386429 500 g 500g
11396429 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11386429 Supplier Thermo Scientific Alfa Aesar Supplier No. 011592.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 7782-85-6 | H15Na2O11P | 268.06 g/mol

Sodium hydrogen phosphate is widely used in detergents and cleaning agents. Combined with trisodium phosphate, it is employed in the food industry to adjust the pH and in water treatment to prevent calcium scale formation. It is utilized as a saline laxative to clean the bowel before a colonoscopy. It prevents the coagulation of condensed milk. It is also utilized as anti-caking additive in powdered products. It enhances the cook time and used as thickening agent in desserts and puddings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-85-6
Molecular Formula H15Na2O11P
Molecular Weight (g/mol) 268.06
MDL Number MFCD00149180
InChI Key LVXHNCUCBXIIPE-UHFFFAOYSA-L
Synonym sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
PubChem CID 6096963
IUPAC Name disodium phosphoric acid heptahydrate
SMILES O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

Melting Point 48.1°C-5H2O
Color White
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 98.0-102.0%
Solubility Information Soluble in water and insoluble in ethanol.
Formula Weight 268.07 (141.98 Anhydrous)
Grade ACS
Odor Odorless
Appearance White crystals
Density 1.7 g/cm3 at 20°C
Chemical Name or Material Sodium hydrogen phosphate heptahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.