missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrogen phosphate, anhydrous, ACS, 99.0% min

Product Code. 11396768
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11396768 5 kg 5kg
11316778 250 g 250g
11306778 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11396768 Supplier Thermo Scientific Alfa Aesar Supplier No. 013437.A7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7558-79-4 | HNa2O4P | 141.96 g/mol

Sodium hydrogen phosphate is used in food and water treatment in conjunction with trisodium phosphate. It is used as an emulsifier and buffer in foods and pharmaceuticals, anti-caking additive in powdered products, dietary supplement, scale inhibitor, reagent and buffer in analytical chemistry. It finds application in detergents, cleaning agents, textile, leather auxiliary, fertilizers and galvanoplastics. Further, it is used as a saline laxative to treat constipation. It inhibits the calcium scale formation in water treatment plants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
IUPAC Name disodium;hydrogen phosphate
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Color White
Physical Form Granules
Quantity 5 kg
Assay Percent Range 99.0% min.
Solubility Information Soluble in water. Insoluble in ethanol,methanol and n-octanol.
Formula Weight 141.98
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Appearance White granular
Chemical Name or Material Sodium hydrogen phosphate, anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.