missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrogen L-tartrate, anhydrous, 98%

Product Code. 11333176
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11333176 500 g 500g
11313176 25 g 25g
11323176 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11333176 Supplier Thermo Scientific Alfa Aesar Supplier No. L12469.36
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 526-94-3 | C4H5NaO6 | 172.07 g/mol

Sodium hydrogen L-tartrate is used for detecting potassium and in nutrient media. It also used as a food additive and an acidity regulator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hydrogen L-tartrate is used for detecting potassium and in nutrient media. It also used as a food additive and an acidity regulator.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 526-94-3
Molecular Formula C4H5NaO6
Molecular Weight (g/mol) 172.07
MDL Number MFCD00065393,MFCD00065393
InChI Key NKAAEMMYHLFEFN-UHFFFAOYNA-M
Synonym sodium bitartrate, monosodium tartrate, sodium hydrogen tartrate, natriumtartrat german, sodium 3-carboxy-2,3-dihydroxypropanoate, monobasic sodium tartrate, monosodium l-+-tartrate, tartaric acid, monosodium salt, natrium hydrogen-2r,3r-tartrat, weinstein
PubChem CID 23690454
IUPAC Name sodium;2,3,4-trihydroxy-4-oxobutanoate
SMILES [Na+].OC(C(O)C([O-])=O)C(O)=O

Specifications

Melting Point ∽243°C (decomposition)
Quantity 500 g
Beilstein 6099125
Merck Index 14,8589
Solubility Information Soluble in water.
Optical Rotation +24.5° (c=3 in Water)
Formula Weight 172.07
Percent Purity 98%
Chemical Name or Material Sodium hydrogen L-tartrate, anhydrous

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.