missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrogen fumarate, 98%

Product Code. 11430558
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430558 250 g 250g
11420558 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430558 Supplier Thermo Scientific Alfa Aesar Supplier No. B24683.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5873-57-4 | C4H2NaO4 | 137.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5873-57-4
Molecular Formula C4H2NaO4
Molecular Weight (g/mol) 137.05
MDL Number MFCD00039097
InChI Key VRVKOZSIJXBAJG-TYYBGVCCSA-L
Synonym sodium hydrogen fumarate, monosodium fumarate, sodium fumarate, maleic acid, sodium salt, maleinian sodu polish, fumaric acid monosodium salt, ccris 7315, 2-butenedioic acid 2z-, sodium salt, sodium e-3-carboxyacrylate, maleinian sodu
PubChem CID 6076814
IUPAC Name sodium;(E)-4-hydroxy-4-oxobut-2-enoate
SMILES [Na+].[O-]C(=O)\C=C\C([O-])=O

Specifications

Quantity 250 g
Formula Weight 138.06
Percent Purity 98%
Chemical Name or Material Sodium hydrogen fumarate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.