missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrogen carbonate, Puratronic™, 99.998% (metals basis)

Product Code. 11307027
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11307027

Brand: Thermo Scientific Alfa Aesar 010863.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

m.p. approx. 50°C, begins to lose CO2 approx.100°C, converts to Na2CO3 | CAS: 144-55-8 | CHNaO3 | 84.01 g/mol

Sodium hydrogen carbonate is used in a process for removing paint and corrosion called sodablasting; the process is particularly suitable for cleaning aluminum panels which can be distorted by other types of abrasive. It can be administered to pools, spas, and garden ponds to raise pH levels. Also can be used as a fire extinguisher. Its primary use is in cooking (baking), as a leavening agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
SMILES [Na+].OC([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.998% (metals basis)
Beilstein 4153970
Solubility Information Soluble in water. Insoluble in ethanol.
IUPAC Name sodium;hydrogen carbonate
Formula Weight 84.01
Odor Odorless
Refractive Index 1.5
Appearance White powder
Density 2.159 g/cm3 at 20°C
Chemical Name or Material Sodium hydrogen carbonate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.