missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrogen arsenate heptahydrate, ACS, 98.0-102.0%

Product Code. p-4407894
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327698 25 g 25g
11317698 100 g 100g
11307698 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11327698 Supplier Thermo Scientific Alfa Aesar Supplier No. 033373.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 10048-95-0 | AsH15Na2O11 | 312.01 g/mol

Sodium hydrogen arsenate heptahydrate is used as a source of soluble arsenic and in dyeing and printing with Turkey-red oil. It also acts as a veterinary drug for anti-parasitic, skin diseases, blood diseases, rheumatism and asthma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10048-95-0
Molecular Formula AsH15Na2O11
Molecular Weight (g/mol) 312.01
MDL Number MFCD00149179
InChI Key KOLXPEJIBITWIQ-UHFFFAOYSA-L
Synonym disodium arsenate heptahydrate, sodium acid arsenate, heptahydrate, sodium arsenate heptahydrate, sodium orthoarsenate heptahydrate, dibasic sodium arsenate heptahydrate, sodium arsenate, dibasic, heptahydrate, unii-90zq7g407w, disodium arsenate, heptahydrate, ccris 5553, arsenic acid h3aso4 , disodium salt, heptahydrate
PubChem CID 61460
SMILES O.O.O.O.O.O.O.[Na+].[Na+].O[As]([O-])([O-])=O

Specifications

Melting Point 130°C (decomposition)
Color White
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 98.0-102.0%
Solubility Information Soluble in water and glycerol.
IUPAC Name disodium heptahydrate hydrogen arsorate
Formula Weight 312.01 (185.94 Anhydrous)
Grade ACS
Appearance White granular
Density 1.88 g/cm3 at 20°C
Chemical Name or Material Sodium hydrogen arsenate heptahydrate, For analysis ACS
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium hydrogen arsenate heptahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
Hazard Statement
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H350-May cause cancer.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Disodium hydrogen arsenate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.