missing translation for 'onlineSavingsMsg'
Learn More

Sodium hyaluronate, 95%

Product Code. 10134623 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10134623 5 g Glass bottle 5g
10491281 1 g Glass bottle 1g
11771371 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10134623 Supplier Thermo Scientific Acros Supplier No. 251770050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9067-32-7 | (C14H20NO11Na)n | 417.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9067-32-7
Molecular Formula (C14H20NO11Na)n
Molecular Weight (g/mol) 417.30
MDL Number MFCD00875848
InChI Key YWIVKILSMZOHHF-QJZPQSOGSA-N
Synonym hyaluronate tetrasaccharide, d0e9sz, 6-3-acetamido-2-6-3-acetamido-2,5-dihydroxy-6-hydroxymethyl oxan-4-yl oxy-2-carboxy-4,5-dihydroxyoxan-3-yl oxy-5-hydroxy-6-hydroxymethyl oxan-4-yl oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
IUPAC Name Sodium hyaluronate
SMILES CC(=O)NC1C(OC2C(O)C(O)C(O-*)OC2C(=O)O[Na])OC(CO)C(O)C1O-*

Specifications

Color White to Cream
pH 6.0 to 8.0 (0.5% aq. soln.)
Loss on Drying 10% max. (500mg, 105°C, 6 hrs)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (On dry substance)
Packaging Glass bottle
Linear Formula (C14H20NO11Na)n
Quantity 5 g
Merck Index 15,4796
Solubility Information Solubility in water: soluble.
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Sodium hyaluronate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.