missing translation for 'onlineSavingsMsg'
Learn More

Sodium hippurate, 96%, Thermo Scientific Chemicals

Product Code. 11439243
Click to view available options
Quantity:
50 g
250 g

It may be a useful urinary biomarker for certain cancers such as gastric cancer and oxidative stress.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It may be a useful urinary biomarker for certain cancers such as gastric cancer and oxidative stress.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 532-94-5
Molecular Formula C9H8NNaO3
Molecular Weight (g/mol) 201.157
MDL Number MFCD00002693
InChI Key ZBCAZEFVTIBZJS-UHFFFAOYSA-M
Synonym sodium hippurate, hippuric acid sodium salt, sodium 2-benzamidoacetate, n-benzoylglycine sodium salt, glycine, n-benzoyl-, monosodium salt, ccris 5812, hippuric acid, monosodium salt, glycine, n-benzoyl-, sodium salt 1:1, hippurate sodium, benzoylaminoacetic acid sodium salt
PubChem CID 516953
IUPAC Name sodium;2-benzamidoacetate
SMILES C1=CC=C(C=C1)C(=O)NCC(=O)[O-].[Na+]

Specifications

Assay Percent Range 96%
Quantity 250 g
Solubility Information Soluble in water.
Formula Weight 201.16
Percent Purity 96%
Physical Form Powder or Lumps
Chemical Name or Material Sodium hippurate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'