missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexanitritocobaltate(III), ACS

Product Code. 11311788
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11311788 100 g 100g
11301788 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11311788 Supplier Thermo Scientific Alfa Aesar Supplier No. 011667.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14649-73-1 | CoN6Na3O12 | 403.933 g/mol

Sodium hexanitritocobaltate(III) is used as primary and secondary intermediate in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14649-73-1
Molecular Formula CoN6Na3O12
Molecular Weight (g/mol) 403.933
MDL Number MFCD00003512
InChI Key ZEBNDJQGEZXBCR-UHFFFAOYSA-H
Synonym cobaltic trisodium hexanitrite, trisodium cobalt 3+ hexanitrite, sodium hexanitritocobaltate 3-, cobalt 3+ sodium nitrite 1:3:6, cobaltate 3-, hexanitrito-, trisodium, cobaltate 3-, hexakis nitrito-o-, trisodium, oc-6-11, na3 co no2 6, cobaltate 3-, hexakis nitrito-kappao-, sodium 1:3 , oc-6-11
PubChem CID 159748
IUPAC Name trisodium;cobalt(3+);hexanitrite
SMILES N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].[Na+].[Na+].[Na+].[Co+3]

Specifications

Melting Point 220°C (decomposition)
Color Yellow to Orange
Physical Form Powder
Quantity 100 g
UN Number UN1479
Merck Index 14,8603
Solubility Information Very soluble in water
Formula Weight 403.94
Grade ACS
Odor Odorless
Chemical Name or Material Sodium hexanitritocobaltate(III)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.