missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexanitritocobaltate(III), ACS

Product Code. 11301788
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11301788

Brand: Thermo Scientific Alfa Aesar 011667.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14649-73-1 | CoN6Na3O12 | 403.933 g/mol

Sodium hexanitritocobaltate(III) is used as primary and secondary intermediate in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14649-73-1
Molecular Formula CoN6Na3O12
Molecular Weight (g/mol) 403.933
MDL Number MFCD00003512
InChI Key ZEBNDJQGEZXBCR-UHFFFAOYSA-H
Synonym cobaltic trisodium hexanitrite, trisodium cobalt 3+ hexanitrite, sodium hexanitritocobaltate 3-, cobalt 3+ sodium nitrite 1:3:6, cobaltate 3-, hexanitrito-, trisodium, cobaltate 3-, hexakis nitrito-o-, trisodium, oc-6-11, na3 co no2 6, cobaltate 3-, hexakis nitrito-kappao-, sodium 1:3 , oc-6-11
PubChem CID 159748
IUPAC Name trisodium;cobalt(3+);hexanitrite
SMILES N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].[Na+].[Na+].[Na+].[Co+3]

Specifications

Melting Point 220°C (decomposition)
Color Yellow to Orange
Physical Form Powder
Quantity 500 g
UN Number UN1479
Merck Index 14,8603
Solubility Information Very soluble in water
Formula Weight 403.94
Grade ACS
Odor Odorless
Chemical Name or Material Sodium hexanitritocobaltate(III)
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.