missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hexametaphosphate, TRC
Specifications
Specifications
| Chemical Name or Material | Sodium Hexametaphosphate |
| CAS | 68915-31-1 |
| Quantity | 10 g |
| Molecular Formula | (NaPO3)n |
| Synonym | Sodium Polyphosphate, Sodium Polyphosphates, Actophos, Brifisol 512, Brifisol 85 Instant, Budit 4, Budit 6H, Budit 7, Budit 9, Calgon, Calgon (old), Calgon 188N, Calgon N, Calgon N New, Calgon R, Calgon Regular, Calgon S, Calgon T, Calgon Unadjusted, Chemi-Carl, Chimotan P, Curafos, Curafos 11-2, Curafos 22-4, Curaphos 700, Dispersant 5010, E 452i, Giltex, Glass H, Glassy Sodium Polyphosphate, Hagan phosphate, Hamine, Hexaphos, Hypersperse SI 300, KW 2353, LSN 3, Medi-Calgon, Micromet, P 8510, Pescamine 150, Poly-phos, Polyrinsan, R-TEK, Rhodiaphos, SHMP, Rumford Metafos, Rumford Quadrafos, S-Tarinsan, SHMP, SQ Phosphate, Sodaphos, Sodium polyphosphate glass, Ultrapolyhos, Vitrafos |
| InChI Formula | InChI=1S/2Na.H4O7P2/c;;1-8(2,3)7-9(4,5)6/h;;(H2,1,2,3)(H2,4,5,6)/q2*+1;/p-2 |
| SMILES | OP(=O)([O-])OP(=O)(O)[O-].[Na+].[Na+] |
| Molecular Weight (g/mol) | 221.94 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?