missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hexametaphosphate, TRC

Product Code. 30396242
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396242 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396242 Supplier Toronto Research Chemicals Supplier No. TRCH6891505G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Hexametaphosphate
CAS 68915-31-1
Quantity 5 g
Molecular Formula (NaPO3)n
Synonym Sodium Polyphosphate, Sodium Polyphosphates, Actophos, Brifisol 512, Brifisol 85 Instant, Budit 4, Budit 6H, Budit 7, Budit 9, Calgon, Calgon (old), Calgon 188N, Calgon N, Calgon N New, Calgon R, Calgon Regular, Calgon S, Calgon T, Calgon Unadjusted, Chemi-Carl, Chimotan P, Curafos, Curafos 11-2, Curafos 22-4, Curaphos 700, Dispersant 5010, E 452i, Giltex, Glass H, Glassy Sodium Polyphosphate, Hagan phosphate, Hamine, Hexaphos, Hypersperse SI 300, KW 2353, LSN 3, Medi-Calgon, Micromet, P 8510, Pescamine 150, Poly-phos, Polyrinsan, R-TEK, Rhodiaphos, SHMP, Rumford Metafos, Rumford Quadrafos, S-Tarinsan, SHMP, SQ Phosphate, Sodaphos, Sodium polyphosphate glass, Ultrapolyhos, Vitrafos
InChI Formula InChI=1S/2Na.H4O7P2/c;;1-8(2,3)7-9(4,5)6/h;;(H2,1,2,3)(H2,4,5,6)/q2*+1;/p-2
SMILES OP(=O)([O-])OP(=O)(O)[O-].[Na+].[Na+]
Molecular Weight (g/mol) 221.94
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.