missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexametaphosphate, tech.

Product Code. p-4407891
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 11327018

Brand: Thermo Scientific Alfa Aesar 014706.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 10124-56-8 | Na6O18P6

Sodium hexametaphosphate is a food additive used in dairy product, canned milk, packaged egg whites, ice cream, seafood and meat processing. It is also used as a sequestrant and water softening agent and detergents. It is an active ingredient in toothpastes as an anti-staining agent. It prevents the steel corrosion. Further, it is used in building, water treatment and in photographic developing. In addition, it is used as a retarder in dental alginate impression materials and as a thinner for petroleum drilling fluids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10124-56-8
Molecular Formula Na6O18P6
MDL Number MFCD00136045
Synonym Sodium metaphosphate

Specifications

Boiling Point 1500°C
Melting Point 628°C
Color White
Physical Form Granules
Quantity 1 kg
Solubility Information Soluble in water. Insoluble in organic solvents
Formula Weight 611.77
Grade Technical
Odor Odorless
Refractive Index 1.482
Appearance White granules
Chemical Name or Material Sodium hexametaphosphate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.