missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexafluoroarsenate(V), 99%, Thermo Scientific Chemicals

Product Code. 11319289
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319289 10 g 10g
11309289 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11319289 Supplier Thermo Scientific Alfa Aesar Supplier No. 088517.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Sodium hexafluoroarsenate(V) is generally used for pharmaceutical intermediates and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12005-86-6
Molecular Formula AsF6Na
Molecular Weight (g/mol) 211.902
MDL Number MFCD00014228
InChI Key NFXMAZFYHDSPPB-UHFFFAOYSA-N
Synonym sodium hexafluoroarsenate v, potassium hexafluoro-$l^ 5-arsanuide, sodium hexafluoro-??-arsanuide, sodium hexafluoroarsenate, naasf6
PubChem CID 14205726
IUPAC Name sodium;hexafluoroarsenic(1-)
SMILES F[As-](F)(F)(F)(F)F.[Na+]

Specifications

Physical Form Powder
Quantity 10 g
UN Number UN1557
Solubility Information Soluble in water.
Formula Weight 211.9
Percent Purity 99%
Chemical Name or Material Sodium hexafluoroarsenate (V)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.