missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexafluoroaluminate, 99.4% (metals basis)

Product Code. 11393428
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11393428

Brand: Thermo Scientific Alfa Aesar 036345.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13775-53-6

Mainly used as fluxing agent in the production of aluminum electrolysis, also used as additive to the wear-resisting of abrasive products, iron alloy and the flux of rimming steel, non-ferrous metal flux, casting deoxidizer, enamel of emulsifier, glass opal agent, welding flux and ceramics filler. Aluminum- metallurgy. This chemical is used as adhesives, cosolvent in enamel industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13775-53-6
MDL Number MFCD00003507
Synonym Aluminum sodium fluoride; Cryolite

Specifications

Melting Point 1,000°C
Physical Form Lump
Quantity 50 g
UN Number UN3260
Solubility Information Sparingly soluble in water 0.4g/L @ 20°C.
Formula Weight 209.95
Percent Purity 99.5%
Odor Odorless
Density 2.90 g/mL
Assay (metals basis)
Chemical Name or Material Sodium hexafluoroaluminate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium hexafluoroaluminate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H332-Harmful if inhaled.
  • H372-Causes damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Sodium hexafluoroaluminate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.