missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexafluoroaluminate, 97.8+%, Thermo Scientific Chemicals

Product Code. 11339229
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339229 500 g 500g
11329229 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339229 Supplier Thermo Scientific Alfa Aesar Supplier No. 088275.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium hexafluoroaluminate is a biochemical for proteomics research. It is used as a flux agent or bath material by aluminum smelters in the production of aluminum. It is also used in the production of synthetic resins for abrasives and in the manufacture of cutting or grinding discs. Minor uses are as a coloring agent (opacifier) in the glass, ceramic and pyrotechnic industries. It occurs naturally as the mineral cryolite, which is used extensively in the industrial production of aluminum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13775-53-6
Molecular Formula Na3AlF6
MDL Number MFCD00003507
Synonym Cryolite; Aluminum sodium fluoride

Specifications

Melting Point 1,000°C
Physical Form Powder
Quantity 500 g
UN Number UN3260
Solubility Information Sparingly soluble in water(0.602g/L at 20°C).
Formula Weight 209.95
Percent Purity ≥97.8%
Odor Odorless
Density 2.90 g/mL
Chemical Name or Material Sodium hexafluoroaluminate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.