missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexacyanoferrate(II) decahydrate, 99%

Product Code. 11394607
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394607 250 g 250g
11384607 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 11394607 Supplier Thermo Scientific Alfa Aesar Supplier No. 039660.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

CAS: 14434-22-1 | C6H20FeN6Na4O10 | 484.06 g/mol

Mainly used as important raw material in pharmaceuticals, pigment, tannage, metallurgy and chemical industries. Used in producing iron blue and potassium ferricyanide, surface corrosion protecting for tannage and metal, carburization treating for carbon steel, removing iron in pharmaceutical producing. Used as oxidant, food additive, explosive and chemical reagent. Also used as additive for preventing agglomeration in melt snow in winter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14434-22-1
Molecular Formula C6H20FeN6Na4O10
Molecular Weight (g/mol) 484.06
MDL Number MFCD00149274
InChI Key JFSUDVTVQZUDOP-UHFFFAOYSA-N
Synonym Sodium ferrocyanide decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[Fe++].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N

Specifications

Melting Point 82°C-10H2O
Color Yellow
Physical Form Powder
Quantity 250 g
Assay Percent Range 99%
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 484.07 (303.92 Anhydrous)
Odor Odorless
Appearance Light yellow powder
Density 1.458
Chemical Name or Material Sodium hexacyanoferrate(II) decahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.