missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexachlororhodate(III) dodecahydrate, Rh 17.1%, Thermo Scientific Chemicals

Catalog Number p-7048871
Click to view available options
Quantity:
1 g
5 g
14972-70-4
Cl6H24Na3O12Rh
600.75
MFCD00003509
JCMYDKHYUGZPJG-UHFFFAOYSA-H
trisodium hexachlororhodiumheptakis(ylium) dodecahydrate
O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].Cl[Rh+7](Cl)(Cl)(Cl)(Cl)Cl
14972-70-4
Cl6H24Na3O12Rh
600.75
MFCD00003509
JCMYDKHYUGZPJG-UHFFFAOYSA-H
trisodium hexachlororhodiumheptakis(ylium) dodecahydrate
O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].Cl[Rh+7](Cl)(Cl)(Cl)(Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 900°C (decomposition)
Quantity 1 g
Formula Weight 600.77 (384.59 Anhydrous)
Concentration or Composition (by Analyte or Components) Rh 17.1%
Physical Form Crystalline
Chemical Name or Material Sodium hexachlororhodate(III) dodecahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'