missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hexachloroplatinate(IV) Hexahydrate, TRC

Product Code. 30421310
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30421310 2.5 g
30415764 1 g
30424638 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30421310 Supplier Toronto Research Chemicals Supplier No. TRCS0772102.5G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19583-77-8
Molecular Formula Cl6Pt • 2Na • 6H2O
Molecular Weight (g/mol) 561.88
Synonym Platinate(2-), hexachloro-, disodium, hexahydrate
IUPAC Name sodium hexachloroplatinate(IV) hexahydrate
SMILES [Cl-][Pt+4]([Cl-])([Cl-])([Cl-])([Cl-])[Cl-].[Na+].O.O.O.O.O.[Na+].O

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/6ClH.2Na.6H2O.Pt/h6*1H;;;6*1H2;/q;;;;;;2*+1;;;;;;;+4/p-6
Formula Weight 558.82
Chemical Name or Material Sodium Hexachloroplatinate(IV) Hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.