missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexachloroplatinate(IV) hexahydrate

Product Code. 11371688
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11371688 5 g 5g
11361688 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11371688 Supplier Thermo Scientific Alfa Aesar Supplier No. 011047.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use As a Catalyst | CAS: 19583-77-8 | Cl6H12Na2O6Pt | 561.85 g/mol

Catalyst.Sodium hexachloroplatinate(IV) hexahydrate is used as a catalyst. It acts as a source of platinum in the preparation of platinum complex. It is also used in the synthesis of platinum nanoparticles using glutathione as a reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19583-77-8
Molecular Formula Cl6H12Na2O6Pt
Molecular Weight (g/mol) 561.85
MDL Number MFCD00149172
InChI Key XGFSWVNCHVSCSP-UHFFFAOYSA-H
Synonym sodium hexachloroplatinate iv hexahydrate, cl6pt.2na.6h2o, platinum iv sodium chloride, disodium;hexachloroplatinum 2-;hexahydrate, disodium hexachloroplatinate 2-hexahydrate
PubChem CID 24846071
SMILES O.O.O.O.O.O.[Na+].[Na+].Cl[Pt](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Melting Point 110°C-6H2O
Color Orange
Physical Form Crystalline
Quantity 5 g
Solubility Information Soluble in water,alcohol. Insoluble in ether.
IUPAC Name disodium hexachloroplatinum hexahydrate
Formula Weight 561.88 (453.79 Anhydrous)
Odor Odorless
Appearance Orange powder
Density 2.5
Chemical Name or Material Sodium hexachloroplatinate(IV) hexahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.