missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexachloroplatinate(IV) hexahydrate, 98%

Product Code. 10606301 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10606301 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10606301 Supplier Thermo Scientific Acros Supplier No. 316750010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 19583-77-8 | Cl6H12Na2O6Pt | 561.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 19583-77-8
Color Orange
Physical Form Crystalline Solid
Quantity 1 g
Assay Percent Range 98%
Molecular Formula Cl6H12Na2O6Pt
Linear Formula Na2PtCl6·6H2O
MDL Number MFCD00149172
Synonym sodium hexachloroplatinate iv hexahydrate, cl6pt.2na.6h2o, platinum iv sodium chloride, disodium;hexachloroplatinum 2-;hexahydrate, disodium hexachloroplatinate 2-hexahydrate
Solubility Information Solubility in water: soluble in water
InChI Key XGFSWVNCHVSCSP-UHFFFAOYSA-H
SMILES O.O.O.O.O.O.[Na+].[Na+].Cl[Pt](Cl)(Cl)(Cl)(Cl)Cl
IUPAC Name disodium hexachloroplatinum hexahydrate
Molecular Weight (g/mol) 561.85
PubChem CID 24846071
Formula Weight 561.88
Percent Purity 98%
Packaging Glass bottle
Chemical Name or Material Sodium hexachloroplatinate(IV) hexahydrate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Sodium hexachloroplatinate(IV) hexahydrate
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: Sodium hexachloroplatinate(IV) hexahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.