missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexachloropalladate(IV) hydrate, 99.9% (metals basis)

Product Code. 11301678
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11301678 1 g 1g
11381668 5 g 5g
11391668 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11301678 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' 011039.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 53823-60-2 | Cl6Na2Pd | 365.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53823-60-2
Molecular Formula Cl6Na2Pd
Molecular Weight (g/mol) 365.10
MDL Number MFCD00014235
InChI Key OWJQEDIJRBKDJO-UHFFFAOYSA-H
IUPAC Name disodium hexachloropalladiumbis(ylium)
SMILES [Na+].[Na+].Cl[Pd++](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Color Brown to Red
Linear Formula Na2PdCl6·xH2O
Quantity 1 g
Sensitivity Hygroscopic
Formula Weight 365.10 (Anhydrous)
Percent Purity 99.9%
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Sodium hexachloropalladate(IV) hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.