missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexachloroosmate(IV) dihydrate, Os 38.7% min

Product Code. 11396558
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11396558 1 g 1g
11386558 0.25 g 0.25g
11376558 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11396558 Supplier Thermo Scientific Alfa Aesar Supplier No. 012176.03
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1307-81-9 ∣ Cl6Na2Os ∣ 448.91 g/mol

Sodium hexachloroosmate(IV) dihydrate is used as an intermediate and catalyst in organic synthesis. It is also used to prepare dihydrogen complexes of osmium(II) containing meso-tetraphos-1 ligand.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1307-81-9
Molecular Formula Cl6Na2Os
Molecular Weight (g/mol) 448.91
MDL Number MFCD00014234
InChI Key PVTVRPSPEDISMV-UHFFFAOYSA-H
PubChem CID 44151190
SMILES [Na+].[Na+].Cl[Os--](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Color Brown to Red
Physical Form Crystalline
Quantity 1 g
Odor Odorless
Sensitivity Air & Moisture Sensitive
Linear Formula Na2OsCl6·2H2O
Solubility Information Insoluble in water.
IUPAC Name disodium;hexachloroosmium(2-)
Formula Weight 484.93 (448.90 Anhydrous)
Grade Ungraded
Concentration or Composition (by Analyte or Components) Os 38.7% min.
Chemical Name or Material Sodium hexachloroosmate(IV) dihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.