missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexachloroosmate(IV) dihydrate, Os 38.7% min

Product Code. 11376558
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
This item is not returnable. View return policy

Product Code. 11376558

Brand: Thermo Scientific Alfa Aesar 012176.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1307-81-9 | Cl6Na2Os | 448.91 g/mol

Sodium hexachloroosmate(IV) dihydrate is used as an intermediate and catalyst in organic synthesis. It is also used to prepare dihydrogen complexes of osmium(II) containing meso-tetraphos-1 ligand.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1307-81-9
Molecular Formula Cl6Na2Os
Molecular Weight (g/mol) 448.91
MDL Number MFCD00014234
InChI Key PVTVRPSPEDISMV-UHFFFAOYSA-H
PubChem CID 44151190
SMILES [Na+].[Na+].Cl[Os--](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Color Brown to Red
Physical Form Crystalline
Quantity 5 g
Linear Formula Na2OsCl6·2H2O
Solubility Information Insoluble in water.
IUPAC Name disodium;hexachloroosmium(2-)
Formula Weight 484.93 (448.90 Anhydrous)
Concentration or Composition (by Analyte or Components) Os 38.7% min.
Odor Odorless
Sensitivity Air & Moisture Sensitive
Chemical Name or Material Sodium hexachloroosmate(IV) dihydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.