missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexachloroiridate(IV) hexahydrate, 99.9+%, (trace metal basis)

Product Code. 10124120 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10124120 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10124120 Supplier Thermo Scientific Acros Supplier No. 197570010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 19567-78-3 | Cl6IrNa2·6H2O | 558.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 600.0°C
CAS 19567-78-3
Color Black to Red
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.9+%
Molecular Formula Cl6IrNa2·6H2O
Linear Formula Na2IrCl6·6H2O
MDL Number MFCD00149169
Synonym sodium hexachloroiridate iv hexahydrate, cl6ir.2na.6h2o, sodium hexachloroiridate 2--water 2/1/6, disodium hexachloroiridate 2-hexahydrate
InChI Key QZVJPVJIAOOTJU-UHFFFAOYSA-H
SMILES O.O.O.O.O.O.[Na+].[Na+].Cl[Ir-2](Cl)(Cl)(Cl)(Cl)Cl
IUPAC Name disodium;hexachloroiridium(2-);hexahydrate
Molecular Weight (g/mol) 558.98
PubChem CID 19849276
Formula Weight 558.98
Percent Purity ≥99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Sodium hexachloroiridate(IV) hexahydrate, 99.9+%
Show More Show Less
compliance-icons
Product Identifier
  • Sodium hexachloroiridate(IV) hexahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.