missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexachloroiridate(IV) hexahydrate, Ir 33.9% min

Product Code. p-7048899
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
missing translation for 'unitSize'
1g
5g
This item is not returnable. View return policy

Product Code. 11331988

missing translation for 'mfr': Thermo Scientific Alfa Aesar 011890.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19567-78-3 | Cl6H12IrNa2O6 | 558.987 g/mol

Sodium hexachloroiridate(IV) hexahydrate is used pharmaceutical Intermediates and organic Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19567-78-3
Molecular Formula Cl6H12IrNa2O6
Molecular Weight (g/mol) 558.987
MDL Number MFCD00149169
InChI Key QZVJPVJIAOOTJU-UHFFFAOYSA-H
Synonym sodium hexachloroiridate iv hexahydrate, cl6ir.2na.6h2o, sodium hexachloroiridate 2--water 2/1/6, disodium hexachloroiridate 2-hexahydrate
PubChem CID 19849276
IUPAC Name disodium;hexachloroiridium(2-);hexahydrate
SMILES O.O.O.O.O.O.[Na+].[Na+].Cl[Ir-2](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Melting Point 600°C (dec.)
Color Black-Red
Physical Form Powder
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 558.99 (450.92 Anhydrous)
Concentration or Composition (by Analyte or Components) Ir 33.9% min.
Chemical Name or Material Sodium hexachloroiridate(IV) hexahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.