missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexabromoplatinate(IV) hexahydrate

Product Code. 11339359
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339359 1 g 1g
11349359 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339359 Supplier Thermo Scientific Alfa Aesar Supplier No. 088966.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39277-13-9 | Br6Na2Pt | 720.488 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39277-13-9
Molecular Formula Br6Na2Pt
Molecular Weight (g/mol) 720.488
MDL Number MFCD00014237
InChI Key RFZKZIKSQSDBBI-UHFFFAOYSA-H
Synonym disodium hexabromoplatinate, sodium hexabromoplatinate iv, disodium hexabromoplatinumdiuide, platinate 2-, hexabromo-, disodium, oc-6-11
PubChem CID 20474473
IUPAC Name disodium;hexabromoplatinum(2-)
SMILES [Na+].[Na+].Br[Pt-2](Br)(Br)(Br)(Br)Br

Specifications

Physical Form Crystalline
Quantity 1 g
Formula Weight 828.49 (720.49 Anhydrous)
Chemical Name or Material Sodium hexabromoplatinate (IV) hexahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.