missing translation for 'onlineSavingsMsg'
Learn More

Sodium Heptanesulfonate, MP Biomedicals

Product Code. 11465144 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465144 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11465144 Supplier MP Biomedicals Supplier No. 0522686701
 Restricted Item
Estimated Shipment: 08-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 22767-50-6
Molecular Formula C7H15NaO3S
Quantity 1 g
Synonym sodium 1-heptanesulfonate, sodium heptane-1-sulfonate, 1-heptanesulfonic acid sodium salt, 1-heptanesulfonic acid, sodium salt, sodium heptane-1-sulphonate, 1-heptanesulfonic acid, sodium salt 1:1, 1-heptanesulfonic acid sodium, sodium heptanesulfonate, 1-heptylsodiumsulfonate, sodium heptane sulfonate
InChI Key REFMEZARFCPESH-UHFFFAOYSA-M
SMILES CCCCCCCS(=O)(=O)[O-].[Na+]
IUPAC Name sodium;heptane-1-sulfonate
Molecular Weight (g/mol) 202.244
PubChem CID 23672332
Chemical Name or Material Sodium Heptane Sulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.