missing translation for 'onlineSavingsMsg'
Learn More

Sodium gualenate, MedChemExpress

Product Code. 16488866
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488866 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16488866 Supplier Medchem Express Supplier No. HYB2191
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sodium gualenate (Guaiazulenesulfonate sodium) is a hydrophilic derivative of guaiazulene with excellent anti-inflammatory and wound-healing effects mainly used for the treatment of duodenal ulcer, gastric ulcer and gastritis.

Sodium gualenate (Guaiazulenesulfonate sodium) is a hydrophilic derivative of guaiazulene with excellent anti-inflammatory and wound-healing effects mainly used for the treatment of duodenal ulcer, gastric ulcer and gastritis.

Specifications

Chemical Name or Material Sodium gualenate
CAS 6223-35-4
Color Black
Purity Grade Notes Research
Molecular Formula C15H17NaO3S
Quantity 100 mg
Synonym Guaiazulenesulfonate sodium
Solubility Information DMSO : 30 mg/mL (99.88 mM; Need ultrasonic and warming)
SMILES O=S(C1=C2C(C)=CC=C(C(C)C)C=C2C(C)=C1)(O[Na])=O
Molecular Weight (g/mol) 300.35
Formula Weight 300.35
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.