missing translation for 'onlineSavingsMsg'
Learn More

Sodium fumarate, 98%

Product Code. 10457192 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10457192 100 g Glass bottle 100g
10291542 5 g Glass bottle 5g
10772505 500 g Glass bottle 500g
15139233 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10457192 Supplier Thermo Scientific Acros Supplier No. 215531000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 17013-01-3 | C4H2Na2O4 | 160.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17013-01-3
Molecular Formula C4H2Na2O4
Molecular Weight (g/mol) 160.04
MDL Number MFCD00064567
InChI Key VXXVUHAXJHEYFH-SEPHDYHBSA-N
Synonym disodium fumarate, sodium fumarate, sodium fumarate dibasic, fumaric acid, disodium salt, fumaric acid disodium salt, fumaran sodny czech, fumaran sodny, disodium butenedioate, di-sodium fumarate, sodium monofumarate
PubChem CID 87110070
IUPAC Name (E)-but-2-enedioic acid;sodium
SMILES C(=CC(=O)O)C(=O)O.[Na].[Na]

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula NaO2CCH=CHCO2Na
Quantity 100 g
Beilstein 02,74
Merck Index 10,4165
Solubility Information Solubility in water: 228g/L water. Other solubilities: insoluble in ethanol, aceton
Formula Weight 160.04
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Sodium fumarate
Show More Show Less
compliance-icons
Product Identifier
  • Sodium fumarate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Sodium fumarate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.