missing translation for 'onlineSavingsMsg'
Learn More

Sodium Formate Solution, 10mM NaOH in isopropanol/water, Solstice

Product Code. 15657590
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Plastic bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15657590 100 mL Plastic bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15657590 Supplier Honeywell-Fluka Supplier No. 97574100ML
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

suitable for LC-MS, 10 mM NaOH in isopropanol/water 1:1 (+0.2% HCOOH)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 141-53-7
Quantity 100 mL
Molecular Formula CHNaO2
Linear Formula HCOONa
MDL Number MFCD00013101
UN Number NONH for all modes of transport
Beilstein 3723428
Synonym sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744
InChI Key HLBBKKJFGFRGMU-UHFFFAOYSA-M
SMILES C(=O)[O-].[Na+]
IUPAC Name sodium;formate
Molecular Weight (g/mol) 68.007
PubChem CID 2723810
ChEBI CHEBI:62965
Formula Weight 68.01g/mol
Packaging Plastic bottle
Chemical Name or Material Sodium formate solution
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium formate solution
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Propan-2-ol, sodiumhydroxide, Sodium formate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.