Learn More
Sodium Formate, Honeywell Fluka™
Brand: Honeywell Fluka 17841-250G
Code : NEW
Additional Details : CAS Number : 141-53-7 Weight : 0.25000kg
Quantity | 250g |
---|
Chemical Identifiers
For HPLC | |
141-53-7 | |
≥99% (NT) | |
HCOONa | |
Soluble in water 0.1g/mL at 25°C |
3595134 | |
Sodium Formate | |
68.01g/mol | |
MFCD00013101 | |
NONH for all modes of transport |
Specifications
Sodium Formate | |
For HPLC | |
HCOONa | |
MFCD00013101 | |
3595134 | |
250g |
141-53-7 | |
≥99% (NT) | |
68.01g/mol | |
NONH for all modes of transport | |
Soluble in water 0.1g/mL at 25°C |