missing translation for 'onlineSavingsMsg'
Learn More

Sodium formaldehydesulfoxylate Dihydrate, Pure, Fisher Chemical

Product Code. 10528260 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10528260 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10528260 Supplier Fisher Chemical Supplier No. S/4040/53

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 63°C
CAS 6035-47-8
Physical Form Solid
Quantity 500 g
Molecular Formula CH7NaO5S
MDL Number 150598
Synonym sodium hydroxymethanesulfinate dihydrate, sodium formaldehydesulfoxylate dihydrate, methanesulfinic acid, hydroxy-, monosodium salt, dihydrate, sodium formaldehyde sulfoxylate, sodium dihydrate hydroxymethanesulfinate, sodium formaldehyde sulfoxylate nf, formaldehyde sodium sulfoxylate dihydrate, monosodium hydroxymethanesulfinate dihydrate, 79-25-4 parent, hydroxymethanesulfinic acid monosodium salt dihydrate
InChI Key UCWBKJOCRGQBNW-UHFFFAOYSA-M
SMILES C(O)S(=O)[O-].O.O.[Na+]
IUPAC Name sodium;hydroxymethanesulfinate;dihydrate
Molecular Weight (g/mol) 154.112
PubChem CID 23666330
Formula Weight 154.11g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Sodium formaldehydesulfoxylate dihydrate
Signal Word
  • Warning
Hazard Category
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H361d-Suspected of damaging the unborn child.
  • H341-Suspected of causing genetic defects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.