missing translation for 'onlineSavingsMsg'
Learn More

Sodium fluorophosphate

Product Code. 11387727
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387727 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11387727 Supplier Thermo Scientific Alfa Aesar Supplier No. 014129.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 10163-15-2 | FNa2O3P | 143.949 g/mol

Sodium fluorophosphate is used as an anti-inflammatory, glucocorticoid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 10163-15-2
Physical Form Powder
Quantity 500 g
Molecular Formula FNa2O3P
MDL Number MFCD00014248
UN Number UN3288
Synonym sodium monofluorophosphate, disodium monofluorophosphate, sodium fluorophosphate, disodium fluorophosphate, phosphorofluoridic acid, disodium salt, sodium phosphorofluoridate, disodium phosphorofluoridate, extra-strength aim, sodium fluorophosphate na2po3f, unii-c810jcz56q
Solubility Information Soluble in water. Insoluble in ethanol and ether.
InChI Key BFDWBSRJQZPEEB-UHFFFAOYSA-L
SMILES [O-]P(=O)([O-])F.[Na+].[Na+]
IUPAC Name disodium;fluoro-dioxido-oxo-$l^{5}-phosphane
Molecular Weight (g/mol) 143.949
PubChem CID 24266
ChEBI CHEBI:86431
Formula Weight 143.95
Odor Odorless
Chemical Name or Material Sodium fluorophosphate
Show More Show Less
compliance-icons
Product Identifier
  • Sodium fluorophosphate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Disodium fluorophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.