missing translation for 'onlineSavingsMsg'
Learn More

Sodium Fluorophosphate, TRC

Código de producto. 30426687
Cambiar vista
Click to view available options
Quantity:
10 g
Tamaño de la unidad:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Tamaño de la unidad
30426687 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30426687 Proveedor Toronto Research Chemicals N.º de proveedor TRCS62750010G
 Restricted Item
Estimated Shipment: 10-12-2026
Añadir a la cesta
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Monofluorophosphate
CAS 10163-15-2
Quantity 10 g
Molecular Formula F O3 P . 2 Na
Synonym Phosphorofluoridic acid','disodium salt (8CI,9CI), Sodium phosphorofluoridate ((NaO)2FPO) (6CI,7CI), Disodium fluorophosphate, Disodium monofluorophosphate, Disodium phosphorofluoridate, Phoskadent Na 211, Sensodyne-Fluor, Sodium fluorophosphate (Na2PFO3), Sodium fluorophosphate (Na2PO3F), Sodium phosphorofluoridate (Na2PO3F)
InChI Formula InChI=1S/FH2O3P.2Na/c1-5(2,3)4;;/h(H2,2,3,4);;/q;2*+1/p-2
SMILES [Na+].[Na+].[O-]P(=O)([O-])F
IUPAC Name disodium;fluoro-dioxido-oxo-λ^{5}-phosphane
Molecular Weight (g/mol) 143.95
Formula Weight 143.9364
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.