missing translation for 'onlineSavingsMsg'
Learn More

Sodium Fluoride, TRC

Product Code. 30411880
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30411880 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30411880 Supplier Toronto Research Chemicals Supplier No. TRCS627000100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Fluoride
CAS 7681-49-4
Quantity 100 g
Molecular Formula F . Na
Synonym Sodium Fluoride, Act, Act (mouthwash), Antibulit, Apoflux, Chemifluor, Dentalfluoro, Duofluorid, Duofluorid F, Duraphat, FDA 0101, Flor-Opal, Floridine, Florinse, Florocid, Fludent, Fluocal, Fluoraday, Fluorigard, Fluorol, Fluorzoin, Fluossen, Flura Drops, Flurexal, Flursol, Flutop Gel, Fungol B, Fuoros, Karidium, Karigel N, Lemoflur, Luride SF, Minute-Gel, Miranol, NSC 77385, Neosten, Neutra-Care, Ora-Bliss, Ossalin, Ossin, Osteo F, Osteofluor, Osteoflur, Pediaflor, Pergantene, Prevident, Prevident 5000 Plus, Prodent, Slow-Fluoride, Sodium monofluoride, Sodium monofluoride (NaF), T-Fluoride, Thera Flur, Wako 194-01971, Winterfresh Gel, Zymafluor
InChI Formula InChI=1S/FH.Na/h1H;/q;+1/p-1
SMILES [F-].[Na+]
IUPAC Name sodium;fluoride
Molecular Weight (g/mol) 41.99
Formula Weight 41.9882
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.