missing translation for 'onlineSavingsMsg'
Learn More

Sodium Fluoride, puriss., Honeywell Fluka™

Product Code. p-7071693 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
1 kg
6 x 1 kg
Unit Size:
1kg
500g
Pack of 6
This item is not returnable. View return policy

Product Code. 15610550

Brand: Honeywell-Fluka 01148500G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, Ph. Eur., USP, 98.5-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
Synonym sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
PubChem CID 5235
ChEBI CHEBI:28741
SMILES [F-].[Na+]

Specifications

Quantity 500 g
Linear Formula NaF
UN Number 1690
IUPAC Name sodium;fluoride
Formula Weight 41.99g/mol
Percent Purity 98.5 to 100.5% (calc. to the dried substance)
Grade puriss.
Vapor Pressure 1.4mmHg (0°C)
Chemical Name or Material Sodium Fluoride, Meets analytical specification of Ph. Eur. BP, USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.