missing translation for 'onlineSavingsMsg'
Learn More

Sodium Fluoride, Mikromol™

Product Code. 30051050
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051050 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30051050 Supplier Mikromol Supplier No. MM0734.00
 Restricted Item
Estimated Shipment: 16-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Sodium Fluoride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Fluoride
CAS 7681-49-4
Analyte or Component Names Sodium Fluoride
Shipping Condition Room Temperature
Molecular Formula F . Na
Certifications/Compliance ISO 17034
Synonym Sodium Fluoride, Act, Act (mouthwash), Antibulit, Apoflux, Chemifluor, Dentalfluoro, Duofluorid, Duofluorid F, Duraphat, FDA 0101, Flor-Opal, Floridine, Florinse, Florocid, Fludent, Fluocal, Fluoraday, Fluorigard, Fluorol, Fluorzoin, Fluossen, Flura Drops, Flurexal, Flursol, Flutop Gel, Fungol B, Fuoros, Karidium, Karigel N, Lemoflur, Luride SF, Minute-Gel, Miranol, NSC 77385, Neosten, Neutra-Care, Ora-Bliss, Ossalin, Ossin, Osteo F, Osteofluor, Osteoflur, Pediaflor, Pergantene, Prevident, Prevident 5000 Plus, Prodent, Slow-Fluoride, Sodium monofluoride, Sodium monofluoride (NaF), T-Fluoride, Thera Flur, Wako 194-01971, Winterfresh Gel, Zymafluor
InChI Formula InChI=1S/FH.Na/h1H;/q;+1/p-1
SMILES [F-].[Na+]
IUPAC Name sodium;fluoride
Molecular Weight (g/mol) 41.99
Quantity 500 mg
Formula Weight 41.9882 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.