missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Fluoride, ACS Reagent, ≥99%, Honeywell Fluka™
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Boiling Point | 1704°C |
Melting Point | 990°C |
Color | Colorless |
pH | 8 to 10 (at 20°C) |
Physical Form | Solid |
Quantity | 100 g |
Linear Formula | NaF |
UN Number | UN1690 |
IUPAC Name | sodium;fluoride |
Formula Weight | 41.99g/mol |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'