missing translation for 'onlineSavingsMsg'
Learn More

Sodium Fluoride, ACS Reagent, ≥99%, Honeywell Fluka™

Catalog Number p-7120789
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7681-49-4
FNa
41.99
MFCD00003524
PUZPDOWCWNUUKD-UHFFFAOYSA-M
sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
5235
CHEBI:28741
[F-].[Na+]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7681-49-4
FNa
41.99
MFCD00003524
PUZPDOWCWNUUKD-UHFFFAOYSA-M
sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
5235
CHEBI:28741
[F-].[Na+]

ACS Reagent,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Boiling Point 1704°C
Melting Point 990°C
Color Colorless
pH 8 to 10 (at 20°C)
Physical Form Solid
Quantity 5 g
Linear Formula NaF
UN Number UN1690
IUPAC Name sodium;fluoride
Formula Weight 41.99g/mol
Percent Purity ≥99%
Grade ACS Reagent
Packaging Poly Bottle
Vapor Pressure 1.4mmHg (0°C)
Density 2.7g/cu.cm
Chemical Name or Material Sodium Fluoride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'