missing translation for 'onlineSavingsMsg'
Learn More

Sodium fluoride, 99%, Thermo Scientific Chemicals

Product Code. 11439933
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11439933 250 g 250g
11429933 1000 g 1000g
11449933 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11439933 Supplier Thermo Scientific Alfa Aesar Supplier No. A13019.30
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Sodium fluoride is used as a steel degreasing agent and in tin electroplating, flux, heat treating salt compositions, fluoridation of drinking water, removing HF from exhaust systems to reduce air pollution, and frosting glasses. It serves as a source of fluoride ion in several applications including it use in synthetic chemistry, for example in Finkelstein reaction for preparating fluoro compounds, desilylation in sensitive organic compounds, metallurgy, and reaction with electrophilic chlorides. It has medical and other applications including complexing of magnesium ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications

Sodium fluoride is used as a steel degreasing agent and in tin electroplating, flux, heat treating salt compositions, fluoridation of drinking water, removing HF from exhaust systems to reduce air pollution, and frosting glasses. It serves as a source of fluoride ion in several applications including it use in synthetic chemistry, for example in Finkelstein reaction for preparating fluoro compounds, desilylation in sensitive organic compounds, metallurgy, and reaction with electrophilic chlorides. It has medical and other applications including complexing of magnesium ions.

Solubility

Slightly soluble in water. Insoluble in alcohol

Notes

It must be kept away from oxidizing agents, metals, acids, alkalis. It is less hygroscopic.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
Synonym sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
PubChem CID 5235
ChEBI CHEBI:28741
SMILES [F-].[Na+]

Specifications

Boiling Point 1704°C
Melting Point 993°C
Physical Form Powder
Quantity 250 g
Linear Formula NaF
UN Number UN1690
Merck Index 14,8618
Solubility Information Slightly soluble in water. Insoluble in alcohol
IUPAC Name sodium fluoride
Formula Weight 41.99
Percent Purity 99%
Grade Ungraded
Odor Odorless
Refractive Index 1.336
Density 2.78
Chemical Name or Material Sodium fluoride
Show More Show Less
compliance-icons
Product Identifier
  • Sodium fluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Sodium fluoride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.