missing translation for 'onlineSavingsMsg'
Learn More

Sodium Fluoride, ≥99%, Reagent Grade, Honeywell™

Product Code. 15678070 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15678070 100 g Poly bottle 100g
15688070 500 g Poly bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15678070 Supplier Honeywell Research Chemicals Supplier No. S1504100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
Synonym sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
PubChem CID 5235
ChEBI CHEBI:28741
IUPAC Name sodium;fluoride
SMILES [F-].[Na+]

Specifications

Boiling Point 1704°C (1013hPa)
Melting Point 990°C
Color Colorless
pH 8.0 to 10.0 (at 20°C)
Physical Form Solid
Quantity 100 g
Linear Formula NaF
UN Number UN1690
Formula Weight 41.99g/mol
Percent Purity ≥99%
Odor Odorless
Packaging Poly bottle
Vapor Pressure 1.4mmHg (0°C)
Density 2.7g/cu.cm at 20°C
Chemical Name or Material Sodium fluoride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.