missing translation for 'onlineSavingsMsg'
Learn More

Sodium Fluoride, ≥99%, (ACS Reagent Grade), MP Biomedicals

Product Code. 11496651 Shop All MP Biomedicals Products
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11496651 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11496651 Supplier MP Biomedicals Supplier No. 0219486480
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 7681-49-4
Quantity 100 g
Assay Percent Range ≥99%
Molecular Formula FNa
MDL Number MFCD00003524
Synonym sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
SMILES [F-].[Na+]
IUPAC Name sodium fluoride
Molecular Weight (g/mol) 41.99
PubChem CID 5235
ChEBI CHEBI:28741
Formula Weight 41.99
Grade ACS Reagent
Chemical Name or Material Sodium Fluoride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.