missing translation for 'onlineSavingsMsg'
Learn More

Sodium Ferric EDDHA, Technical Grade, TRC

Product Code. 30384737
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384737 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384737 Supplier Toronto Research Chemicals Supplier No. TRCE33510350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Ferric EDDHA
CAS 16455-61-1
Molecular Formula C18 H16 N2 O6 . Fe . Na
Quantity 50 mg
Synonym Abreverd Fe-PM, Chelafer, Dissolvine Q, Dissolvine Q 15, Dissolvine Q 40
InChI Formula InChI=1S/C18H20N2O6.Fe.Na/c21-13-7-3-1-5-11(13)15(17(23)24)19-9-10-20-16(18(25)26)12-6-2-4-8-14(12)22;;/h1-8,15-16,19-22H,9-10H2,(H,23,24)(H,25,26);;/q;+3;+1/p-4
SMILES [Na+].[Fe+3].[O-]C(=O)C(NCCNC(C(=O)[O-])c1ccccc1[O-])c2ccccc2[O-]
IUPAC Name sodium;2-[2-[[carboxylato-(2-oxidophenyl)methyl]amino]ethylamino]-2-(2-oxidophenyl)acetate;iron(3+)
Molecular Weight (g/mol) 435.164
Formula Weight 435.026
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.