missing translation for 'onlineSavingsMsg'
Learn More

Sodium Ethoxide (>85%), TRC

Product Code. 30419961
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
30419961 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 30419961

Brand: Toronto Research Chemicals TRCS62500025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Ethoxide
CAS 141-52-6
Molecular Formula C2H5NaO
Quantity 25 g
Synonym Ethanol Sodium Salt, Ethyl Alcohol Sodium Salt, Caustic Alcohol, Ethoxysodium, Sodium Ethanolate, Sodium Ethoxylate, Sodium Ethylate
InChI Formula InChI=1S/C2H5O.Na/c1-2-3;/h2H2,1H3;/q-1;+1
SMILES [Na+].CC[O-]
IUPAC Name sodium;ethanolate
Molecular Weight (g/mol) 68.05
Formula Weight 68.0238
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.